Learn More
Thermo Scientific™ KingFisher™ Systems Consumables
Catalog No. 22387023 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
KingFisher plate 100 μL
KingFisher plate 200 μL
KingFisher tip comb
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-387-023 KingFisher plate 200 μL
22-387-021 KingFisher tip comb
22-387-022 KingFisher plate 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-387-023 Supplier Thermo Scientific™ Supplier No. 97002084
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart
Optimize performance using Thermo Scientific™ KingFisher™Systems Consumables, ideal for magnetic particle processing.
Optimize performance using Thermo Scientific™ KingFisher™ Systems Consumables, specifically designed for use with the KingFisher System and ideal for magnetic particle processing because of their low binding affinity for biomolecules. The specially-designed, disposable tip combs and microplates, which are made of polypropylene, enable excellent recovery of magnetic beads.
  • Specifically designed for use with the KingFisher System
  • Ideal for magnetic particle processing because of their low binding affinity for biomolecules
  • The specially-designed, disposable tip combs and microplates are made of polypropylene
  • This enables excellent recovery of magnetic beads
TRUSTED_SUSTAINABILITY
Specifications
Capacity 200 μL
For Use With (Application) Magnetic particle processing
For Use With (Equipment) KingFisher System
Includes KingFisher plate 200 μL
Quantity 50/Cs.
Are the MagMAX Express and KingFisher plates the same?
Yes, the MagMAX Express and KingFisher plates are equivalent. We recommend the KingFisher plates as an alternative to the discontinued MagMAX Express plates.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.