Learn More
Grainger Revvo Rigid Plate Caster, Polyurethane, 5 in Wheel, 550 lbs. Capacity, 1 ea
Catalog No. 502790549
Encompass_Preferred
These rigid casters feature a fabricated steel frame for enhanced durability and tracking in light-to-medium-duty applications. Designed for industrial use, this caster includes a high-performance polyurethane wheel that provides a smooth, quiet roll on a variety of surfaces.
  • Rigid caster configuration for stable tracking
  • Durable 12.7 cm (5 in.) diameter polyurethane wheel
  • Zinc plated steel frame for corrosion resistance
  • Load capacity of 249.5 kg (550 lbs.)
  • Plain bore wheel bearing for maintenance-free operation
  • Operating temperature range from -30°C to +80°C (-22°F to +176°F)
  • Non-marking tread construction
Catalog No. 50-279-0549 Supplier Grainger Supplier No. 16D130
These rigid casters feature a fabricated steel frame for enhanced durability and tracking in light-to-medium-duty applications. Designed for industrial use, this caster includes a high-performance polyurethane wheel that provides a smooth, quiet roll on a variety of surfaces.
  • Rigid caster configuration for stable tracking
  • Durable 12.7 cm (5 in.) diameter polyurethane wheel
  • Zinc plated steel frame for corrosion resistance
  • Load capacity of 249.5 kg (550 lbs.)
  • Plain bore wheel bearing for maintenance-free operation
  • Operating temperature range from -30°C to +80°C (-22°F to +176°F)
  • Non-marking tread construction