Learn More
Grainger Approved Vendor Swivel Plate Caster, Polyurethane, 8 in. Wheel, 1,200 lb. Capacity, 1 ea
Catalog No. 502792448
Encompass_Preferred
These kingpinless plate casters feature an 8 in. diameter, debris-resistant polyurethane wheel designed for quiet, smooth mobility in diverse industrial environments. With an extended lead for reduced starting effort, these casters enhance tracking and minimize chatter to decrease structural stress and extend service life.
  • Kingpinless swivel construction for durability
  • 87 Shore A wheel hardness for floor protection
  • 1,200 lb. load capacity suitable for medium-duty applications
  • Donut-shaped tread design
  • Zinc-plated aluminum frame finish
  • Suitable for surfaces including asphalt, ceramic, grease, oil, steel, and wood
  • Integrated ball bearings for smooth rotation
Catalog No. 50-279-2448 Supplier Grainger Supplier No. 19YW86
These kingpinless plate casters feature an 8 in. diameter, debris-resistant polyurethane wheel designed for quiet, smooth mobility in diverse industrial environments. With an extended lead for reduced starting effort, these casters enhance tracking and minimize chatter to decrease structural stress and extend service life.
  • Kingpinless swivel construction for durability
  • 87 Shore A wheel hardness for floor protection
  • 1,200 lb. load capacity suitable for medium-duty applications
  • Donut-shaped tread design
  • Zinc-plated aluminum frame finish
  • Suitable for surfaces including asphalt, ceramic, grease, oil, steel, and wood
  • Integrated ball bearings for smooth rotation