Learn More
Grainger Zoro Select Kingpinless Swivel Plate Caster, Polyurethane, 8 in, 1,200 lb, 1 ea
Catalog No. 502792608
Encompass_Preferred
This kingpinless swivel plate caster is designed for heavy-duty material handling, featuring a durable polyurethane wheel with an aluminum core for reliable performance. Engineered for demanding industrial environments, this caster delivers superior mobility and durability.
  • Load capacity of 1,200 lb
  • Kingpinless swivel raceway for increased durability and reduced maintenance
  • Precision ball bearings ensure smooth, quiet operation
  • Non-marking polyurethane tread protects floors
  • Debris-resistant design ideal for diverse surface types including wood, metal, and tile
  • Zinc-plated steel frame provides excellent corrosion resistance
Catalog No. 50-279-2608 Supplier Grainger Supplier No. 19YW82
This kingpinless swivel plate caster is designed for heavy-duty material handling, featuring a durable polyurethane wheel with an aluminum core for reliable performance. Engineered for demanding industrial environments, this caster delivers superior mobility and durability.
  • Load capacity of 1,200 lb
  • Kingpinless swivel raceway for increased durability and reduced maintenance
  • Precision ball bearings ensure smooth, quiet operation
  • Non-marking polyurethane tread protects floors
  • Debris-resistant design ideal for diverse surface types including wood, metal, and tile
  • Zinc-plated steel frame provides excellent corrosion resistance