Learn More
Athens Research And Technology Inc KINNINOGEN HMW HU 100UG
Supplier:  Athens Research And Technology Inc 1616110914H100UG
Encompass_Preferred
Kininogen, High Molecular Weight, Human Plasma: 100 UG; Frozen in 10 mM Na acetate, pH 5.5, with 250 mM NaCl; MW 120,000; EC= 0.70; Purity: > =95% by SDS-PAGE. Native; Purified from human plasma in the Athens Research & Tech lab.
Catalog No. 50-489-834