Learn More
Cayman Chemical KIntensInhumntrIfluoro 10mg
SDP
Catalog No. 502950942
Encompass_Preferred
A neurotensin ligand; binds to neurotensin receptors in HT-29 cells and mouse brain (Kis = 1,396 and 149.6 nM, respectively) but does not induce the accumulation of intracellular calcium in HT-29 cells (EC50 = >10,000 nM); induces histamine release in isolated rat peritoneal mast cells (EC50 = ~10 µM); induces vascular permeability in rats
Catalog No. 50-295-0942 Supplier Cayman Chemical Supplier No. 3313810 MG
A neurotensin ligand; binds to neurotensin receptors in HT-29 cells and mouse brain (Kis = 1,396 and 149.6 nM, respectively) but does not induce the accumulation of intracellular calcium in HT-29 cells (EC50 = >10,000 nM); induces histamine release in isolated rat peritoneal mast cells (EC50 = ~10 µM); induces vascular permeability in rats