Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kinyoun's Carbol-Fuchsin Solution, Sigma-Aldrich

Catalog No. 01258989 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-989 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-989 Supplier MilliporeSigma Supelco Supplier No. 21819250MLF
Add to Cart
Add to Cart

Kinyoun's carbol-fuchsin is intended for use to stain paraffin sections of a formaldehyde-fixed specimen containing tubercle bacilli, and for the demonstration of tuberculosis bacteria (Mycobacterium tuberculosis). It is also applicable for other organisms that retain the dye after immersion in acidified 70% alcohol. It has been used to develop a staining technique for detecting microsporidial spores in fecal specimens.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbol-Fuchsin solution according to Kinyoun
Color Dark Red
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.