Learn More
Electron Microscopy Sciences Kinyoun's Solution 1 LT
SDP
Catalog No. 5026758
Encompass_Preferred
C₂₁H₂₂ClN₃
Kinyoun's carbol fuchsin solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It is a staining solution for acid-fast bacteria.
Catalog No. 50-267-58 Supplier Electron Microscopy Sciences Supplier No. 2608706
C₂₁H₂₂ClN₃
Kinyoun's carbol fuchsin solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It is a staining solution for acid-fast bacteria.