Learn More
Electron Microscopy Sciences Kinyoun's Solution 500 ML
SDP
Catalog No. 5026759
Encompass_Preferred
C₂₁H₂₂ClN₃
Kinyoun's carbol fuchsin solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It is a staining solution for acid-fast bacteria.
Catalog No. 50-267-59 Supplier Electron Microscopy Sciences Supplier No. 2608705
C₂₁H₂₂ClN₃
Kinyoun's carbol fuchsin solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It is a staining solution for acid-fast bacteria.