Learn More
Aviva Systems Biology Corp KIR2DL1 Antibody - C-terminal region (ARP63462_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63462_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIR2DL1. It was validated on Western Blot. . Alternate names: CD158A, KIR-K64, KIR221, NKAT, NKAT1, p58.1ApplicationIP, WB. Species reactivity: Human
Catalog No. 50-287-974