Learn More
Medchemexpress LLC KIR2DL1 Human Biot 20ug | 20ug
SDP
Catalog No. 5000193820
Encompass_Preferred
KIR2DL1 found on NK cells serves as a receptor for specific HLA-C alleles inhibiting NK cell activity to prevent cell lysis This regulatory function involves interactions with ARRB2 and additionally engages PTPN6 and PTPN11 The interaction s enhancement by ARRB2 underscores KIR2DL1 s role in complex signaling networks that modulate NK cell function KIR2DL1 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived KIR2DL1 protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-93820 Supplier Medchemexpress LLC Supplier No. HYP7816520UG
KIR2DL1 found on NK cells serves as a receptor for specific HLA-C alleles inhibiting NK cell activity to prevent cell lysis This regulatory function involves interactions with ARRB2 and additionally engages PTPN6 and PTPN11 The interaction s enhancement by ARRB2 underscores KIR2DL1 s role in complex signaling networks that modulate NK cell function KIR2DL1 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived KIR2DL1 protein expressed by HEK293 with C-Avi C-His labeled tag