Learn More
Aviva Systems Biology Corp KIR2DL5A Antibody - C-terminal region (ARP65773_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65773_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIR2DL5A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human KIR2DL5A. Alternate names: CD158F, KIR2DL5, KIR2DL5.1, KIR2DL5.3ApplicationWB. Species reactivity: Human
Catalog No. 50-288-568