Learn More
Aviva Systems Biology Corp KIR2DS2 Antibody - N-terminal region (ARP63891_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63891_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIR2DS2. It was validated on Western Blot. . Alternate names: CU464060.1, 183ActI, CD158J, CD158b, NKAT5, cl-49ApplicationWB. Species reactivity: Human
Catalog No. 50-288-108