Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci KIR2DS2 Peptide
SDP

Catalog No. 16595983
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-983 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-983 Supplier ProSci Supplier No. 8201P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

KIR2DS2 peptide is used for blocking the activity of KIR2DS2 antibody.

Specifications

Gene ID (Entrez) 100132285
Species Human
Accession Number NP_036444
Common Name Killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 2
Concentration 200 μg/mL
Content And Storage Store KIR2DS2 peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol KIR2DS2
Quantity 0.05 mg
Sequence 18 amino acid peptide near the carboxy terminus of human KIR2DS2.
Synonym Killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 2, KIR2DL3, MHC class I NK cell receptor, Natural killer-associated transcript 5, NKAT5, p58 KIR, p58 natural killer cell receptor clone CL-49, p58 NK receptor
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.