Learn More
Selleck Chemical LLC kira6 5mg 1589527-65-0
SDP
Supplier:  Selleck Chemical LLC S86585MG
Encompass_Preferred
kira6 is a potent type II IRE1? kinase inhibitor with an IC50 of 0.6 ?M. It dose-dependently inhibits IRE1?(WT) kinase activity, XBP1 RNA cleavage, Ins2 RNA cleavage and oligomerization. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8144