Learn More
Medchemexpress LLC HY-114368 5mg Medchemexpress, Kira8 CAS:1630086-20-2 Purity:>98%
SDP
Catalog No. 501932483
Encompass_Preferred
Medchemexpress, HY-114368 5mg Kira8 CAS:1630086-20-2 Kira8 (AMG-18) is a mono-selective IRE1α inhibitor that allosterically attenuates IRE1α RNase activity with an IC 50 of 5.9 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2483 Supplier Medchemexpress LLC Supplier No. HY1143685MG
Medchemexpress, HY-114368 5mg Kira8 CAS:1630086-20-2 Kira8 (AMG-18) is a mono-selective IRE1α inhibitor that allosterically attenuates IRE1α RNase activity with an IC 50 of 5.9 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.