Learn More
AdipoGen Kirromycin
SDP
Supplier:  AdipoGen BVT0157M001
Encompass_Preferred
Chemical. CAS 50935-71-2. Formula C43H60N2O12. MW 796.9. Isolated from Streptomyces collinus. Antibiotic. Member of the elfermycin group. Antibacterial. Protein biosynthesis inhibitor. Specifically interacts with the bacterial elongation factor EF-Tu and prevents the release of EF-Tu from the bacterial ribosome after GTP hydrolysis. In the absence of ribosomes it induces EF-Tu-dependent hydrolysis of GTP. Animal growth promoter used in poultry breeding.
Catalog No. 50-256-2084