Learn More
Abcam Kirromycin, Antibiotic agent, 1MG
Catalog No. 502705920
Encompass_Preferred
MW 796.9 Da, Purity >96%. Potent antibiotic agent. Selective EF-Tu inhibitor. Protein synthesis inhibitor. Prevents EF-Tu release from the ribosome. Induces EF-Tu-dependent hydrolysis of GTP in the absence of ribosomes. Shows potent bactericidal effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5920 Supplier Abcam Supplier No. AB1442871MG
MW 796.9 Da, Purity >96%. Potent antibiotic agent. Selective EF-Tu inhibitor. Protein synthesis inhibitor. Prevents EF-Tu release from the ribosome. Induces EF-Tu-dependent hydrolysis of GTP in the absence of ribosomes. Shows potent bactericidal effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement