Learn More
Cayman Chemical Kisspeptin-234 (trifluoroacetate salt), Purity: ≥98%, CAS: 1848962-29-7, 10 mg
SDP
Catalog No. 502913479
Encompass_Preferred
A peptide kisspeptin receptor antagonist; inhibits kisspeptin-10-induced inositol phosphate production in CHO cells expressing the human kisspeptin receptor (IC50 = 7 nM) and kisspeptin-10-induced firing of isolated mouse GnRH neurons; inhibits pulsatile GnRH secretion in pubertal female rhesus monkeys; inhibits kisspeptin-10-induced luteinizing hormone secretion in adult male rats and mice
Catalog No. 50-291-3479 Supplier Cayman Chemical Supplier No. 3563310 MG
A peptide kisspeptin receptor antagonist; inhibits kisspeptin-10-induced inositol phosphate production in CHO cells expressing the human kisspeptin receptor (IC50 = 7 nM) and kisspeptin-10-induced firing of isolated mouse GnRH neurons; inhibits pulsatile GnRH secretion in pubertal female rhesus monkeys; inhibits kisspeptin-10-induced luteinizing hormone secretion in adult male rats and mice