Learn More
Medchemexpress LLC Kiss2 peptide acetate | 1111070-93-9 | 10 MG
SDP
Catalog No. 5000425508
Encompass_Preferred
Kiss2 peptide acetate is the acetate form of Kiss2 peptide. It is a positive regulator of reproduction, binding to its cognate receptor Kiss2R (GPR54) in COS-7 cells. This activates PKA and PKC signaling pathways through Gas and Gaq proteins, enhancing the activity of cAMP response element-dependent luciferase (CRE-luc) and serum response element-dependent luciferase (SRE-luc).
  • Targets Kisspeptin Receptor, PKA, PKC
  • Involved in GPCR/G Protein, Stem Cell/Wnt, TGF-beta/Smad, Epigenetics pathways
  • Soluble in DMSO at 100 mg/mL, requires ultrasonic
  • Enhances activity of CRE-luc and SRE-luc
  • Activates PKA and PKC signaling pathways through Gas and Gaq proteins
Catalog No. 50-004-25508 Supplier Medchemexpress LLC Supplier No. HYP10256A10MG
Kiss2 peptide acetate is the acetate form of Kiss2 peptide. It is a positive regulator of reproduction, binding to its cognate receptor Kiss2R (GPR54) in COS-7 cells. This activates PKA and PKC signaling pathways through Gas and Gaq proteins, enhancing the activity of cAMP response element-dependent luciferase (CRE-luc) and serum response element-dependent luciferase (SRE-luc).
  • Targets Kisspeptin Receptor, PKA, PKC
  • Involved in GPCR/G Protein, Stem Cell/Wnt, TGF-beta/Smad, Epigenetics pathways
  • Soluble in DMSO at 100 mg/mL, requires ultrasonic
  • Enhances activity of CRE-luc and SRE-luc
  • Activates PKA and PKC signaling pathways through Gas and Gaq proteins