Learn More
CPC Scientific H-Phe-Asn-Phe-Asn-Pro-Phe-Gly-Leu-Arg-Phe-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific KISS005A
Encompass
Kisspeptin peptides and its receptor, GPR54, play key roles in cancer and reproduction development. In mammals (except for the platypus), there is only one kisspeptin gene (Kiss1) present. In fish and some amphibians, two kisspeptin genes (Kiss1 and Kiss2) are common, and have been observed in zebra fish, goldfish, chub mackerel, and sea bass.ONE-LETTER SEQUENCE: FNFNPFGLRF-NH2MOLECULAR FORMULA: C63H84N16O12MOLECULAR WEIGHT:1257.45STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: Beck, Benjamin H., et al. Comparative Biochemistry and Physiology Part A: Molecular & Integrative Physiology 162.3 (2012): 265-273.; Kotani, M. et al. The Journal of Biological chemistry 276, (2001) 34631?34636.; Mechaly, Alejandro S., Jordi Vinas, and Francesc Piferrer. Molecular and Cellular Endocrinology 339, no. 1-2 (2011): 14-24.
Catalog No. 50-210-9623