Learn More
Eurofins Discoverx Kisspeptin-10/Metastin, 1 mg. (92-1121)
Supplier:  Eurofins Discoverx 921121
Encompass
Kisspeptin-10 is a protein and potent endogenous agonist for the kisspeptin receptor, GPR54 (Ki=2.33 nM). Kisspeptin-GPR54 signaling assists in gonadotropin-releasing hormone secretion at puberty. Kisspeptin-10 is the smallest fully bioactive fragment of kisspeptin, and was originally identified as a metastasis suppressor for melanoma and breast cancer. Provided as 1 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9654