Learn More
Medchemexpress LLC Kisspeptin 13 TFA | 98.9% | 1626.81 | 1 MG
SDP
Catalog No. 5000396641
Encompass_Preferred
Kisspeptin 13 TFA is the TFA salt form of Kisspeptin 13 (HY-P3641). It inhibits glucose-induced insulin secretion with an IC50 of 1.2 nM. Kisspeptin 13 TFA activates the hypothalamic-pituitary-adrenal (HPA) axis, causes hyperthermia, motor behavior and anxiety in rats. This compound interacts with α2-adrenergic and 5-HT2 serotonin receptors and exhibits antidepressant efficacy. It is an activator for GPR54 and GnRH receptor, enhancing memory and being useful in Alzheimer's disease research.
  • Inhibits glucose-induced insulin secretion (IC50 of 1.2 nM)
  • Activates hypothalamic-pituitary-adrenal (HPA) axis
  • Causes hyperthermia, motor behavior and anxiety in rats
  • Interacts with α2-adrenergic and 5-HT2 serotonin receptors
  • Exhibits antidepressant efficacy
  • Activator for GPR54 and GnRH receptor
  • Enhances memory
  • Useful in Alzheimer's disease research
Catalog No. 50-003-96641 Supplier Medchemexpress LLC Supplier No. HYP3641A1MG
Kisspeptin 13 TFA is the TFA salt form of Kisspeptin 13 (HY-P3641). It inhibits glucose-induced insulin secretion with an IC50 of 1.2 nM. Kisspeptin 13 TFA activates the hypothalamic-pituitary-adrenal (HPA) axis, causes hyperthermia, motor behavior and anxiety in rats. This compound interacts with α2-adrenergic and 5-HT2 serotonin receptors and exhibits antidepressant efficacy. It is an activator for GPR54 and GnRH receptor, enhancing memory and being useful in Alzheimer's disease research.
  • Inhibits glucose-induced insulin secretion (IC50 of 1.2 nM)
  • Activates hypothalamic-pituitary-adrenal (HPA) axis
  • Causes hyperthermia, motor behavior and anxiety in rats
  • Interacts with α2-adrenergic and 5-HT2 serotonin receptors
  • Exhibits antidepressant efficacy
  • Activator for GPR54 and GnRH receptor
  • Enhances memory
  • Useful in Alzheimer's disease research