Learn More
CPC Scientific H-Ile-Pro-Ala-Pro-Gln-Gly-Ala-Val-Leu-Val-Gln-Arg-Glu-Lys-Asp-Leu-Pro-Asn-Tyr-Asn-Trp-Asn-Ser-Phe-Gly-Leu-Arg-Phe-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific KISS003B
Encompass
An LRF-amide motif containing fragment of malignant melanoma metastasis-suppressor Kiss1 which binds to human GPR54, a G-protein-coupled receptor also known as AXOR12. It shows lower agonistic potency towards AXOR12 than malignant melanoma metastasis-suppressor Kisspeptin-13 (4-13) (human) (H-5544).ONE-LETTER SEQUENCE: IPAPQGAVLVQREKDLPNYNWNSFGLRF-NH2MOLECULAR FORMULA: C149H226N42O39MOLECULAR WEIGHT:3229.69STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1135442-77-1], SYNONYMS: Metastin (27-54) (human), KiSS-1 (94-121) (human), Malignant Melanoma Metastasis-Suppressor KiSS-1 (94-121) (human)RESEARCH AREA: CancerREFERENCES: A.West et al., Genomics, 54, 145 (1998); A.I.Muir et al., J. Biol. Chem., 276, 28969 (2001)
Catalog No. 50-210-9622