Learn More
Waters Corp Quality Parts Kit, 1/16 in. Fitting, Mechanical, UPLC
Catalog No. 502391416 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement kit provides a 1/16 in. fitting set designed for one end of a 1/16 in. delay coil, commonly used with the Fraction Manager Analytical. It ensures proper connection and functionality for your system.
  • Spare parts kits
  • Mechanical type
  • UPLC system compatibility
  • As required replacement schedule
Catalog No. 50-239-1416 Supplier Waters Corp Supplier No. 700011563
This replacement kit provides a 1/16 in. fitting set designed for one end of a 1/16 in. delay coil, commonly used with the Fraction Manager Analytical. It ensures proper connection and functionality for your system.
  • Spare parts kits
  • Mechanical type
  • UPLC system compatibility
  • As required replacement schedule