Learn More
Waters Corp Syringe Kit, 12.5 mL, 10 mL Injection, for 3767 Sample Manager, HPLC
Supplier:  Waters Corp 700013085
Encompass_Preferred
This Syringe Kit enables 10mL injections. It includes the Syringe, Sample, and Holding Loops necessary to support the specified injection volumes on a 3767 Sample Manager.
  • Upgrade kit
  • Designed for HPLC systems
  • Replaced as required
  • Includes syringe, sample, and holding loops
  • Enables 10 mL injections on a 3767 sample manager
Catalog No. 50-239-2739