Learn More
Chemglass Life Sciences Kit, Minum-Ware, Intermediate
SDP
Catalog No. 501532675
Encompass_Preferred
Kit, Minum-Ware, Intermediate. Similar to the MW-11-01 Deluxe MINUM-WARE kit with the exception of the components required to perform the GC experiments described in Organic Laboratory Techiques, Donald L. Pavia, Gary M. Lampman, George S. Kriz, Randy G. Engel, Sanders College Publishing-Chicago IL. and Microscale Organic Laboratory, Dana W. Mayo, Ronald M. Pike, Samuel S. Butcher, John S. Wiley & Sons, NY., NY. Kit also contains one rather than two each of the 3.0 and 5.0ml conical reaction vials. The MW-13-01 kit is packaged in a polypropylene box having movable dividers permitting additional components to be added at a later date. All joints are 14/10 with the outer members externally threaded. Large quantity discounts available, please call for details. For complete list of components, see the specifications tab below.
Catalog No. 50-153-2675 Supplier Chemglass Life Sciences Supplier No. MW1301
Add to Cart
Add to Cart
Kit, Minum-Ware, Intermediate. Similar to the MW-11-01 Deluxe MINUM-WARE kit with the exception of the components required to perform the GC experiments described in Organic Laboratory Techiques, Donald L. Pavia, Gary M. Lampman, George S. Kriz, Randy G. Engel, Sanders College Publishing-Chicago IL. and Microscale Organic Laboratory, Dana W. Mayo, Ronald M. Pike, Samuel S. Butcher, John S. Wiley & Sons, NY., NY. Kit also contains one rather than two each of the 3.0 and 5.0ml conical reaction vials. The MW-13-01 kit is packaged in a polypropylene box having movable dividers permitting additional components to be added at a later date. All joints are 14/10 with the outer members externally threaded. Large quantity discounts available, please call for details. For complete list of components, see the specifications tab below.