Learn More
Waters Corp Syringe Kit, 2.5 mL, 3767 Sample Manager
Catalog No. 502391412 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit is designed for use with the 3767 Sample Manager. It contains components for sample aspiration, dispensing, and injection, including a 2.5 mL syringe, a holding loop, and a sample loop.
  • Includes a 2.5 mL syringe for aspirating and dispensing samples.
  • Features a holding loop that isolates the sample from the syringe.
  • Contains a sample loop that holds the sample for injection.
  • Compatible with the 3767 Sample Manager.
Catalog No. 50-239-1412 Supplier Waters Corp Supplier No. 700013083
This kit is designed for use with the 3767 Sample Manager. It contains components for sample aspiration, dispensing, and injection, including a 2.5 mL syringe, a holding loop, and a sample loop.
  • Includes a 2.5 mL syringe for aspirating and dispensing samples.
  • Features a holding loop that isolates the sample from the syringe.
  • Contains a sample loop that holds the sample for injection.
  • Compatible with the 3767 Sample Manager.