Learn More
Waters Corp Syringe Kit, 25 mL, for 3767 Sample Manager, HPLC
Catalog No. 502392853 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit is an upgrade for the 3767 Sample Manager, designed for HPLC systems. It facilitates 20 mL injections and includes a syringe, sample, and holding loops to support the stated injection volumes.
  • Upgrade kit
  • For HPLC system
  • Allows 20 mL injections
  • Contains syringe, sample, and holding loops
  • Designed for use with the 3767 Sample Manager
  • Replacement schedule as required
Catalog No. 50-239-2853 Supplier Waters Corp Supplier No. 700013086
This kit is an upgrade for the 3767 Sample Manager, designed for HPLC systems. It facilitates 20 mL injections and includes a syringe, sample, and holding loops to support the stated injection volumes.
  • Upgrade kit
  • For HPLC system
  • Allows 20 mL injections
  • Contains syringe, sample, and holding loops
  • Designed for use with the 3767 Sample Manager
  • Replacement schedule as required