Learn More
Waters Corp Syringe Kit, 5.0 mL, for 3767 Sample Manager, HPLC
Catalog No. 502392472 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This syringe kit allows for 5 mL injections, providing the necessary components to support stated injection volumes on a 3767 Sample Manager.
  • Contains syringe, sample, and holding loops
  • Allows for 5 mL injections
  • Functions as an upgrade kit
  • Compatible with HPLC systems
  • Replaced as needed for optimal performance
  • Provided as a spare parts kit
Catalog No. 50-239-2472 Supplier Waters Corp Supplier No. 700013084
This syringe kit allows for 5 mL injections, providing the necessary components to support stated injection volumes on a 3767 Sample Manager.
  • Contains syringe, sample, and holding loops
  • Allows for 5 mL injections
  • Functions as an upgrade kit
  • Compatible with HPLC systems
  • Replaced as needed for optimal performance
  • Provided as a spare parts kit