Learn More
Newcomer Supply Alcian Blue/PAS Stain Kit, 250 ml kit
SDP
Catalog No. NC1677419
Encompass
Newcomer Supply Alcian Blue/PAS Stain Kit procedure is used to differentiate acidic epithelial mucins (sialomucin, sulfomucin), neutral epithelial mucins and stromal (mesenchymal) mucins and is a means of detecting the overall presence of mucins.
Catalog No. NC1677419 Supplier Newcomer Supply Supplier No. 91022A
Newcomer Supply Alcian Blue/PAS Stain Kit procedure is used to differentiate acidic epithelial mucins (sialomucin, sulfomucin), neutral epithelial mucins and stromal (mesenchymal) mucins and is a means of detecting the overall presence of mucins.