Learn More
Chemglass Life Sciences Kit, Amber 2.0mL 12x32mm 9mm Thread Large Opening Vial and a 0.040" Red PTFE/Silicone Septa Lined BONDED CLOSURE
SDP
Catalog No. 501532636
Encompass_Preferred
Kit, Amber 2.0mL 12x32mm 9mm Thread Large Opening Vial and a 0.040" Red PTFE/Silicone Septa Lined BONDED CLOSURE. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.
Catalog No. 50-153-2636 Supplier Chemglass Life Sciences Supplier No. CV51721232
Kit, Amber 2.0mL 12x32mm 9mm Thread Large Opening Vial and a 0.040" Red PTFE/Silicone Septa Lined BONDED CLOSURE. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.