Learn More
Beckman Coulter Beckman Coulter Multisizer 4e Aperture Tube Kit, 50 μm (C93975)
Catalog No. NC3199647
Encompass_Preferred
The Beckman Coulter Multisizer 4e Aperture Tube kit provides a 50 μm sensing aperture for replacement and particle size and distribution analysis on Multisizer instruments. It is intended for use with Multisizer 4 and 4e systems and supplies a ready-to-install aperture tube for routine maintenance.
  • Aperture size: 50 μm
  • Particle size analysis range: 1-30 μm
  • Compatible with Multisizer 4 and 4e instruments only
  • Intended for particle size and distribution analysis
  • Supplied as a replacement aperture kit for routine maintenance
Catalog No. NC3199647 Supplier Beckman Coulter Supplier No. C93975
The Beckman Coulter Multisizer 4e Aperture Tube kit provides a 50 μm sensing aperture for replacement and particle size and distribution analysis on Multisizer instruments. It is intended for use with Multisizer 4 and 4e systems and supplies a ready-to-install aperture tube for routine maintenance.
  • Aperture size: 50 μm
  • Particle size analysis range: 1-30 μm
  • Compatible with Multisizer 4 and 4e instruments only
  • Intended for particle size and distribution analysis
  • Supplied as a replacement aperture kit for routine maintenance