Learn More
Molecular Devices Molecular Devices Reporter Assay Kit, Firefly Luciferase, Glow, Explorer
Catalog No. 5000014603
Encompass_Preferred
The SpectraMax Glo Steady-Luc Reporter Assay Kit provides a sensitive, glow-type luminescence assay for quantifying firefly luciferase expression in mammalian cells. It offers extended signal stability and a wide dynamic range suitable for high-throughput microplate screening workflows.
  • Sensitive quantitation of firefly luciferase in mammalian cells.
  • Glow-type luminescence with extended signal stability up to 5 hours (within 20% of initial signal).
  • Linear detection across approximately seven orders of magnitude.
  • Explorer kit sufficient for 2 x 96-well plates; bulk option available for higher throughput.
  • Optimized for luminescence microplate readers and plate reader software workflows.
  • Suitable for high-throughput screening and batch plate processing.
Catalog No. 50-000-14603 Supplier Molecular Devices Supplier No. R8352
The SpectraMax Glo Steady-Luc Reporter Assay Kit provides a sensitive, glow-type luminescence assay for quantifying firefly luciferase expression in mammalian cells. It offers extended signal stability and a wide dynamic range suitable for high-throughput microplate screening workflows.
  • Sensitive quantitation of firefly luciferase in mammalian cells.
  • Glow-type luminescence with extended signal stability up to 5 hours (within 20% of initial signal).
  • Linear detection across approximately seven orders of magnitude.
  • Explorer kit sufficient for 2 x 96-well plates; bulk option available for higher throughput.
  • Optimized for luminescence microplate readers and plate reader software workflows.
  • Suitable for high-throughput screening and batch plate processing.