Learn More
Molecular Devices Molecular Devices IMAP FP Evaluation Kit, Progressive Binding, 384-Well
Catalog No. 5000014387
Encompass_Preferred
The IMAP FP Evaluation Kit provides reagents for fluorescence polarization (FP) assays to evaluate kinase and phosphatase activity using IMAP bead-based binding chemistry. The kit contains proprietary IMAP binding reagent and binding buffers configured to stop enzyme reactions and initiate binding, enabling FP readout in high-throughput plate formats. Labeled substrates are available separately.
  • Provides reagents for FP-based kinase and phosphatase assays.
  • Includes proprietary binding reagent and binding buffers to stop reactions and initiate bead binding.
  • Configured for high-throughput screening in 384-well plates.
  • Sufficient reagents to generate approximately 800 data points in a standard 384-well format.
  • Labeled substrates sold separately for assay customization.
Catalog No. 50-000-14387 Supplier Molecular Devices Supplier No. R8155
The IMAP FP Evaluation Kit provides reagents for fluorescence polarization (FP) assays to evaluate kinase and phosphatase activity using IMAP bead-based binding chemistry. The kit contains proprietary IMAP binding reagent and binding buffers configured to stop enzyme reactions and initiate binding, enabling FP readout in high-throughput plate formats. Labeled substrates are available separately.
  • Provides reagents for FP-based kinase and phosphatase assays.
  • Includes proprietary binding reagent and binding buffers to stop reactions and initiate bead binding.
  • Configured for high-throughput screening in 384-well plates.
  • Sufficient reagents to generate approximately 800 data points in a standard 384-well format.
  • Labeled substrates sold separately for assay customization.