Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Devices Molecular Devices FLIPR Calcium 5 Assay Kit, 100-Plate, 2 Vials

Catalog No. 5000014658
Encompass_Preferred

The FLIPR Calcium 5 Assay Kit is a fluorescence-based, homogeneous reagent kit for measuring intracellular calcium flux in cell-based assays using FLIPR systems. It provides an increased signal window and supports incubation at 37°C or room temperature for high-throughput, automated screening.

  • High signal window for improved assay sensitivity.
  • Suitable for 96-, 384-, and 1536-well plate formats.
  • Contains two reagent vials, each sufficient for approximately 50 plates.
  • Kit sufficient for approximately 100 plates.
  • Compatible with automated and overnight screening workflows.
  • Assay dilution/assay buffer is supplied separately.

Catalog No. 50-000-14658 Supplier Molecular Devices Supplier No. R8187
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The FLIPR Calcium 5 Assay Kit is a fluorescence-based, homogeneous reagent kit for measuring intracellular calcium flux in cell-based assays using FLIPR systems. It provides an increased signal window and supports incubation at 37°C or room temperature for high-throughput, automated screening.

  • High signal window for improved assay sensitivity.
  • Suitable for 96-, 384-, and 1536-well plate formats.
  • Contains two reagent vials, each sufficient for approximately 50 plates.
  • Kit sufficient for approximately 100 plates.
  • Compatible with automated and overnight screening workflows.
  • Assay dilution/assay buffer is supplied separately.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.