Learn More
Med Vet International Open Suction Catheter Kit, 2 Gloves, Delee Tip, 8 Fr, 200 mL Cup, 50/Case, Dynd40978
Catalog No. 502831073
Encompass_Preferred
Medline suction catheters are better by design. Medline carries catheters for every need. From adult to neonatal, DeLee or whistle tips, single catheters or convenient wet and dry kits, we have just the right components for your procedure. Why risk contamination when you can use Medline suction catheters with the exclusive Contro-Vac valve - proven more effective than chimney valves at preventing contact with fluids. Our 5-in-1 Connector and Stabilizing Bar ensure easy connection for any size tubing and a secure grip. In an independent laboratory evaluation, Medline suction catheters with the Contro-Vac valve were compared to leading manufacturer's catheters with chimney valves. The results? Medline catheters were proven more effective at preventing the release of microbial contamination during suctioning process. Catheter, Cup & Gloves 8Fr (DeLee Tip), 2 Gloves.
Catalog No. 50-283-1073 Supplier Med Vet International Supplier No. DYND40978
Medline suction catheters are better by design. Medline carries catheters for every need. From adult to neonatal, DeLee or whistle tips, single catheters or convenient wet and dry kits, we have just the right components for your procedure. Why risk contamination when you can use Medline suction catheters with the exclusive Contro-Vac valve - proven more effective than chimney valves at preventing contact with fluids. Our 5-in-1 Connector and Stabilizing Bar ensure easy connection for any size tubing and a secure grip. In an independent laboratory evaluation, Medline suction catheters with the Contro-Vac valve were compared to leading manufacturer's catheters with chimney valves. The results? Medline catheters were proven more effective at preventing the release of microbial contamination during suctioning process. Catheter, Cup & Gloves 8Fr (DeLee Tip), 2 Gloves.