Learn More
Chemglass Life Sciences Kit, Clear 2.0mL 12x32mm 9mm Thread Large Opening Vial and a PTFE/Silicone Septa Lined BONDED CLOSURE With Slit
SDP
Catalog No. 501532634
Encompass_Preferred
Kit, Clear 2.0mL 12x32mm 9mm Thread Large Opening Vial and a PTFE/Silicone Septa Lined BONDED CLOSURE With Slit. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.
Catalog No. 50-153-2634 Supplier Chemglass Life Sciences Supplier No. CV51691232
Kit, Clear 2.0mL 12x32mm 9mm Thread Large Opening Vial and a PTFE/Silicone Septa Lined BONDED CLOSURE With Slit. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.