Learn More
Carnabio Usa Inc KIT(D816V)/5UG/CYTOPLASMICKIT
Supplier:  Carnabio Usa Inc 085055
Encompass
KIT [D816V], Cytoplasmic domain, Mutant [D816V], Amino Acid 544-976, NP_000213.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0479