Learn More
Invitrogen™ Human KIT [D816V], GST Tag, His Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Human KIT, C-terminal fragment, amino acids T544-V976 (as in NCBI/Protein entry NP_000213.1) with a D816V mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Related Product: LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant
Specifications
Specifications
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Format | Kit |
| Formulation | 50mM HEPES with 5mM DTT, 20% glycerol, 15mM glutathione, 100mM NaCl and no preservative; pH 7.5 |
| Molecular Weight (g/mol) | 77.4 kDa |
| Name | Human KIT [D816V], GST Tag, His Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 1 mg |
| Storage Requirements | -80°C, Avoid Freeze/Thaw Cycles |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.