Learn More
Thermo Scientific Purge Screw Valve
Supplier:  Thermo Scientific 6040.2035
Encompass
Purge screw for mass spectrometry ion source made of stainless steel rated for 100 megapascals of pressure with VF-P20 threading to vent and evacuate gases while maintaining vacuum.
Catalog No. 50-134-7795