Learn More
Chemglass Life Sciences Disposable Tubing Accessory Kit
SDP
Catalog No. 501532102
Encompass
Tubing Kit provides simple options to replace the tubes in the Tubing Complete Set by avoiding a wear out of the tube during operation.It is recommended to replace the tubes after using it three times.
Catalog No. 50-153-2102 Supplier Chemglass Life Sciences Supplier No. CLS133001
Tubing Kit provides simple options to replace the tubes in the Tubing Complete Set by avoiding a wear out of the tube during operation.It is recommended to replace the tubes after using it three times.