Learn More
Med Vet International Jorvet Tracheal Wash Kit, J0283
Catalog No. 5000495336
Encompass_Preferred
Transtracheal wash is a very useful diagnostic tool in the treatment of lower respiratory disease in horses. The JorVet Tracheal Wash kit provides a convenient and easy method to obtain high-quality diagnostic samples. The procedure is simple to perform under light sedation with a small local block over the lower third of the neck. Sterile. 15g cannula and 16g needle Catheter 22"L
Catalog No. 50-004-95336 Supplier Med Vet International Supplier No. JOR283
Transtracheal wash is a very useful diagnostic tool in the treatment of lower respiratory disease in horses. The JorVet Tracheal Wash kit provides a convenient and easy method to obtain high-quality diagnostic samples. The procedure is simple to perform under light sedation with a small local block over the lower third of the neck. Sterile. 15g cannula and 16g needle Catheter 22"L