Learn More
Waters Corp Waters Corporation Ferrule, Back Locking, 1/16 in., PEEK
Catalog No. NC2236841 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This back locking ferrule is a widely used replacement part for various systems.
  • Back locking design
  • Made from PEEK material
  • Suitable for 1/16 in. tubing
  • Units per package: 2
  • Designed for UPLC system type
  • Replacement schedule: As required
This item is not returnable. View return policy
Catalog No. NC2236841 Supplier Waters Corp Supplier No. 700011767
This back locking ferrule is a widely used replacement part for various systems.
  • Back locking design
  • Made from PEEK material
  • Suitable for 1/16 in. tubing
  • Units per package: 2
  • Designed for UPLC system type
  • Replacement schedule: As required