Learn More
Molecular Devices Molecular Devices dsDNA Assay Kit, Fluorescence, 468/507 nm
Catalog No. 5000014356
Encompass_Preferred
The SpectraMax Quant AccuClear Nano dsDNA Assay Kit is a fluorescence-based assay for selective quantitation of double-stranded DNA in 96-well microplate format. It uses a dye that preferentially binds dsDNA and provides a wide linear detection range suitable for low to high DNA concentrations.
  • Excitation wavelength 468 nm, 9 nm bandwidth.
  • Emission wavelength 507 nm, 15 nm bandwidth.
  • Linear detection range 34 pg to 250 ng per well in 96-well format.
  • Bulk kit format sufficient for ten 96-well microplates.
  • Includes 100X dye and 20X buffer plus eight dsDNA standard vials.
Catalog No. 50-000-14356 Supplier Molecular Devices Supplier No. R8357
The SpectraMax Quant AccuClear Nano dsDNA Assay Kit is a fluorescence-based assay for selective quantitation of double-stranded DNA in 96-well microplate format. It uses a dye that preferentially binds dsDNA and provides a wide linear detection range suitable for low to high DNA concentrations.
  • Excitation wavelength 468 nm, 9 nm bandwidth.
  • Emission wavelength 507 nm, 15 nm bandwidth.
  • Linear detection range 34 pg to 250 ng per well in 96-well format.
  • Bulk kit format sufficient for ten 96-well microplates.
  • Includes 100X dye and 20X buffer plus eight dsDNA standard vials.