Learn More
Molecular Devices Molecular Devices dsDNA Assay Kit, Fluorescence, 2-2000 ng, 350/460 nm
Catalog No. 5000014484
Encompass_Preferred
The SpectraMax Quant AccuBlue HiRange dsDNA Assay Kit is a fluorescence-based assay designed for accurate quantitation of concentrated double-stranded DNA samples in a 96-well microplate format. The kit provides dye, buffer, enhancer, and a set of dsDNA standards for a workflow-optimized assay with a broad linear range.
  • Fluorescence excitation and emission at 350 nm and 460 nm respectively.
  • Linear detection range of 2 ng to 2000 ng dsDNA per well in a 96-well plate.
  • Includes concentrated dye, enhancer, buffer, and calibrated dsDNA standards for assay setup.
  • High selectivity for double-stranded DNA over single-stranded nucleic acids and RNA.
  • Preconfigured protocols available for compatible microplate readers and analysis software.
  • For research use only; not for diagnostic procedures.
Catalog No. 50-000-14484 Supplier Molecular Devices Supplier No. R8359
The SpectraMax Quant AccuBlue HiRange dsDNA Assay Kit is a fluorescence-based assay designed for accurate quantitation of concentrated double-stranded DNA samples in a 96-well microplate format. The kit provides dye, buffer, enhancer, and a set of dsDNA standards for a workflow-optimized assay with a broad linear range.
  • Fluorescence excitation and emission at 350 nm and 460 nm respectively.
  • Linear detection range of 2 ng to 2000 ng dsDNA per well in a 96-well plate.
  • Includes concentrated dye, enhancer, buffer, and calibrated dsDNA standards for assay setup.
  • High selectivity for double-stranded DNA over single-stranded nucleic acids and RNA.
  • Preconfigured protocols available for compatible microplate readers and analysis software.
  • For research use only; not for diagnostic procedures.