Learn More
Waters Corp Waters Frit and Nut Kit, Stainless Steel, 0.2 μm, 2.1 mm, 5-pack
Catalog No. 502039451 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides essential replacement components for maintaining optimal performance in UPLC systems. It includes five 0.2 μm stainless steel replacement filters and five replacement inlet nuts, designed for use in conjunction with compatible column in-line filter kits.
  • Contains five 0.2 μm stainless steel replacement filters.
  • Includes five replacement inlet nuts.
  • Designed for use with compatible column in-line filter kits.
  • Functions as a fitting within the system.
  • Compatible with UPLC system types.
  • Constructed from stainless steel for durability.
  • Replacement recommended as required.
  • Provided in a package of five units.
Catalog No. 50-203-9451 Supplier Waters Corp Supplier No. 700002775
This kit provides essential replacement components for maintaining optimal performance in UPLC systems. It includes five 0.2 μm stainless steel replacement filters and five replacement inlet nuts, designed for use in conjunction with compatible column in-line filter kits.
  • Contains five 0.2 μm stainless steel replacement filters.
  • Includes five replacement inlet nuts.
  • Designed for use with compatible column in-line filter kits.
  • Functions as a fitting within the system.
  • Compatible with UPLC system types.
  • Constructed from stainless steel for durability.
  • Replacement recommended as required.
  • Provided in a package of five units.