Learn More
Waters Corp Waters Corporation Vial, LCGC Certified, Clear Glass, 12 x 32 mm, 2 mL
Supplier:  Waters Corp 186000272C
Encompass_Preferred
These LCGC Certified vials are rigorously tested for cleanliness and dimensional tolerances, crucial for chromatographers. Made from chemically inert Type 1, 33 expansion borosilicate glass, their clear design ensures easy visibility.
  • Certified vials are HPLC tested for cleanliness.
  • Vials maintain tight dimensional tolerances for autosamplers.
  • Batches undergo cleanliness testing post-packaging.
  • Headspace GC tests confirm proper silicone septa curing.
  • Universal screw cap provides a superior seal.
  • Paired with PTFE/Silicone septum for multiple injections and storage.
  • PTFE/Silicone septa offer excellent resealing; working temperature -40°C to 200°C.
  • Constructed from Type 1, 33 expansion borosilicate glass.
  • Clear glass allows for easy solution visibility.
Catalog No. 50-818-629