Learn More
BrainBits BrainBits Complete Culturing Kit (Hippocampus)
Catalog No. NC1091410
Encompass
BrainBits Complete Culturing Kit - The BrainBits Complete Culturing Kit contains everything you need to start a culture from fresh embryonic tissue. - This kit is perfect for the researcher just starting out or for those who want reliability. - Great for classroom education allowing a hands-on approach for the next generation researchers. - Tissue pair in 2 mL of Hibernate EB Complete.12 mL of NbActiv1 media for initiating a culture of primary rat or mouse neurons. 6.3 mg of sterile Papain. 5.0 mL of HE-Ca. A 9 silanized, fire polished pipette and bulb. A pair of 50 µg/mL poly-D-lysine coated 15 mm glass coverslips. A primary neuronal culture protocol.
Catalog No. NC1091410 Supplier BrainBits Supplier No. KTC57EHP
BrainBits Complete Culturing Kit - The BrainBits Complete Culturing Kit contains everything you need to start a culture from fresh embryonic tissue. - This kit is perfect for the researcher just starting out or for those who want reliability. - Great for classroom education allowing a hands-on approach for the next generation researchers. - Tissue pair in 2 mL of Hibernate EB Complete.12 mL of NbActiv1 media for initiating a culture of primary rat or mouse neurons. 6.3 mg of sterile Papain. 5.0 mL of HE-Ca. A 9 silanized, fire polished pipette and bulb. A pair of 50 µg/mL poly-D-lysine coated 15 mm glass coverslips. A primary neuronal culture protocol.