Learn More
Waters Corp Kit, I-Class ACQUITY UPLC, PEEKsil Needle
Catalog No. NC2393407 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides a PEEKsil needle for ACQUITY UPLC I-Class systems, serving as a replacement part or accessory for the ACQUITY I-Class PLUS Sample Manager-FL and ACQUITY I-Class Sample Manager-FL.
  • Designed for ACQUITY UPLC I-Class systems
  • Made of PEEKsil material
  • Functions as a replacement part or accessory
  • Compatible with ACQUITY I-Class PLUS Sample Manager-FL
  • Compatible with ACQUITY I-Class Sample Manager-FL
Catalog No. NC2393407 Supplier Waters Corp Supplier No. 700005930
This kit provides a PEEKsil needle for ACQUITY UPLC I-Class systems, serving as a replacement part or accessory for the ACQUITY I-Class PLUS Sample Manager-FL and ACQUITY I-Class Sample Manager-FL.
  • Designed for ACQUITY UPLC I-Class systems
  • Made of PEEKsil material
  • Functions as a replacement part or accessory
  • Compatible with ACQUITY I-Class PLUS Sample Manager-FL
  • Compatible with ACQUITY I-Class Sample Manager-FL